3-Boc-6-oxo-3-aza-bicyclo[3.1.1]heptane 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 6-oxo-3-azabicyclo[3.1.1]heptane-3-carboxylate
A155457-100MG
A155457-250MG
A155457-1G
76984-804EA
151.71
USD
76984-804
77611-478
77683-302
3-Boc-6-oxo-3-aza-bicyclo[3.1.1]heptane 95%
3-Boc-6-oxo-3-aza-bicyclo[3.1.1]heptane
Precaución: For research use only.
Formula:
C₁₁H₁₇NO₃ MW: 211.258 g/mol |
MDL Number:
MFCD17016750 CAS Number: 1251013-26-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...