Morpholine-4-sulfonyl chloride 95%

Supplier: Ambeed
Danger

A155311-1G A155311-5G A155311-25G
77028-348EA 9.55 USD
77028-348 77028-350 77671-196
Morpholine-4-sulfonyl chloride 95%
Morpholine-4-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₄H₈ClNO₃S
MDL Number: MFCD03249981
CAS Number: 1828-66-6
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR