trans-Ethyl 4-aminocyclohexanecarboxylate hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-aminocyclohexane-1-carboxylate, hydrochloride

A155145-100MG A155145-1G A155145-250MG A155145-5G
77010-956EA 5.31 USD
77010-956 77010-958 77010-960 77010-962
trans-Ethyl 4-aminocyclohexanecarboxylate hydrochloride 95%
trans-Ethyl 4-aminocyclohexanecarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₇NO₂·ClH
MW: 207.698 g/mol
MDL Number: MFCD14635812
CAS Number: 2084-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR