6-Fluoro-2-methylnicotinaldehyde 97%

Supplier: Ambeed
Warning

A155100-1G A155100-250MG A155100-100MG A155100-5G
77028-338EA 209 USD
77028-338 77028-340 77871-767 77871-768
6-Fluoro-2-methylnicotinaldehyde 97%
6-Fluoro-2-methylnicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₆FNO
MDL Number: MFCD05662399
CAS Number: 884494-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR