(S)-2-Aminopent-4-ynoic acid hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-Propargyl-Gly-OH·HCl

A155083-10G A155083-250MG A155083-5G
76997-500EA 241.89 USD
76997-500 76997-502 77671-194
(S)-2-Aminopent-4-ynoic acid hydrochloride 97%
(S)-2-Aminopent-4-ynoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₅H₈ClNO₂
MW: 149.58 g/mol
MDL Number: MFCD03659731
CAS Number: 198774-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR