Sulfamethazine sodium salt 98%

Supplier: Ambeed
Warning

A155058-100G A155058-10G A155058-25G A155058-500G A155058-5G
76997-490EA 32.89 USD
76997-490 76997-492 76997-494 76997-496 76997-498
Sulfamethazine sodium salt 98%
Sulfamethazine sodium salt

Precaución: for Research Use Only

Formula: C₁₂H₁₃N₄NaO₂S
MDL Number: MFCD00068333
CAS Number: 1981-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR