(E)-3-(3,5-Dichlorophenyl)acrylic acid 98%

Supplier: Ambeed
Warning

A155021-250MG A155021-1G
76984-788EA 11 USD
76984-788 77885-265
(E)-3-(3,5-Dichlorophenyl)acrylic acid 98%
(E)-3-(3,5-Dichlorophenyl)acrylic acid

Precaución: for Research Use Only

Formula: C₉H₆Cl₂O₂
MDL Number: MFCD00179444
CAS Number: 20595-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR