5-Bromo-7-(trifluoromethyl)-1H-indazole 98%

Supplier: Ambeed
Warning

A154926-250MG A154926-1G
77010-950EA 51.99 USD
77010-950 77683-299
5-Bromo-7-(trifluoromethyl)-1H-indazole 98%
5-Bromo-7-(trifluoromethyl)-1H-indazole

Precaución: For research use only.

Formula: C₈H₄BrF₃N₂
MW: 265.03 g/mol
MDL Number: MFCD21602947
CAS Number: 1374258-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR