2-(3-Fluoropyridin-2-yl)acetonitrile 95%

Supplier: Ambeed
Warning

A154882-1G A154882-250MG
76984-784EA 259.93 USD
76984-784 76984-786
2-(3-Fluoropyridin-2-yl)acetonitrile 95%
2-(3-Fluoropyridin-2-yl)acetonitrile

Precaución: for Research Use Only

Formula: C₇H₅FN₂
MW: 136.126 g/mol
MDL Number: MFCD09924179
CAS Number: 149488-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR