3-Bromo-5-chloro-2-methylaniline 97%

Supplier: Ambeed
Warning

A154867-1G A154867-250MG A154867-25G
77028-334EA 140.04 USD
77028-334 77028-336 77592-902
3-Bromo-5-chloro-2-methylaniline 97%
3-Bromo-5-chloro-2-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇BrClN
MW: 220.4942 g/mol
MDL Number: MFCD12198860
CAS Number: 1166756-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR