Ethyl 4,6-dichloro-2-(methylthio)pyrimidine-5-carboxylate 97%

Supplier: Ambeed
Warning

A154830-100MG A154830-10G A154830-1G A154830-250MG A154830-5G
77028-320EA 42.44 USD
77028-320 77028-322 77028-324 77028-326 77028-328
Ethyl 4,6-dichloro-2-(methylthio)pyrimidine-5-carboxylate 97%
Ethyl 4,6-dichloro-2-(methylthio)pyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₈H₈Cl₂N₂O₂S
MW: 267.13 g/mol
MDL Number: MFCD12828684
CAS Number: 959070-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR