2-(4-Bromophenyl)propanenitrile 95%
Supplier: Ambeed
|
Warning
|
A154698-1G
A154698-250MG
A154698-5G
76984-774EA
121
USD
76984-774
76984-776
76984-778
2-(4-Bromophenyl)propanenitrile 95%
2-(4-Bromophenyl)propanenitrile
Precaución: for Research Use Only
Formula:
C₉H₈BrN |
MDL Number:
MFCD11036513 CAS Number: 42186-06-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...