1-(3-Methoxy-5-methylphenyl)ethanone 95%

Supplier: Ambeed
Warning

A154555-250MG A154555-1G A154555-100MG
77028-308EA 25 USD
77028-308 77600-198 77871-765
1-(3-Methoxy-5-methylphenyl)ethanone 95%
1-(3-Methoxy-5-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MDL Number: MFCD17011766
CAS Number: 43113-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR