Zeatin riboside 96%

Supplier: Ambeed
Warning

Synonyms: trans-Zeatinriboside, trans-Zeatin-riboside

A154489-10MG A154489-1MG
77028-302EA 18 USD
77028-302 77028-304
Zeatin riboside 96%
Zeatin riboside

Precaución: for Research Use Only

Formula: C₁₅H₂₁N₅O₅
MW: 351.363 g/mol
MDL Number: MFCD00036809
CAS Number: 6025-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR