1-Chloro-2,5-difluoro-3-nitrobenzene 95%

Supplier: Ambeed
Warning

A154415-1G A154415-250MG A154415-5G
77010-940EA 280.08 USD
77010-940 77010-942 77010-944
1-Chloro-2,5-difluoro-3-nitrobenzene 95%
1-Chloro-2,5-difluoro-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂ClF₂NO₂
MW: 193.535 g/mol
MDL Number: MFCD27923474
CAS Number: 1805029-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR