Methyl 5-bromo-2-hydroxy-4-(trifluoromethyl)benzoate 98%

Supplier: Ambeed
Warning

A154144-100MG A154144-250MG
76984-766EA 17 USD
76984-766 76984-768
Methyl 5-bromo-2-hydroxy-4-(trifluoromethyl)benzoate 98%
Methyl 5-bromo-2-hydroxy-4-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₆BrF₃O₃
MDL Number: MFCD11110855
CAS Number: 1131587-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR