3-Amino-2-fluoro-4-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4-methylpyridin-3-amine

A154119-100G A154119-1G
76997-474EA 1337.8 USD
76997-474 77671-166
3-Amino-2-fluoro-4-methylpyridine 98%
3-Amino-2-fluoro-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇FN₂
MW: 126.13 g/mol
MDL Number: MFCD04112556
CAS Number: 173435-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR