Methyl 7-hydroxy-2-naphthoate 98%

Supplier: Ambeed
Warning

A154075-100MG A154075-1G A154075-250MG A154075-5G
77010-930EA 27 USD
77010-930 77010-932 77611-462 77892-051
Methyl 7-hydroxy-2-naphthoate 98%
Methyl 7-hydroxy-2-naphthoate

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₃
MW: 202.21 g/mol
MDL Number: MFCD11226994
CAS Number: 95901-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR