5-Bromo-1,3-diethyl-2-iodobenzene 95%

Supplier: Ambeed
Warning

A153935-1G A153935-25G A153935-5G
76984-760EA 22.28 USD
76984-760 76984-762 76984-764
5-Bromo-1,3-diethyl-2-iodobenzene 95%
5-Bromo-1,3-diethyl-2-iodobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrI
MW: 339.01 g/mol
MDL Number: MFCD09743748
CAS Number: 942475-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR