2-Chloro-5-methylpyrimidine-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A153928-100MG A153928-1G A153928-250MG A153928-5G
77028-276EA 14 USD
77028-276 77028-278 77028-280 77028-282
2-Chloro-5-methylpyrimidine-4-carboxylic acid 95%
2-Chloro-5-methylpyrimidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
MDL Number: MFCD13193603
CAS Number: 933746-10-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR