2-Amino-3,5-dibromopyridine 97%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dibromo-2-pyridylamine, 3,5-Dibromopyridin-2-amine

A153927-100G A153927-10G A153927-25G A153927-500G
76984-752EA 35.01 USD
76984-752 76984-754 76984-756 76984-758
2-Amino-3,5-dibromopyridine 97%
2-Amino-3,5-dibromopyridine

Precaución: for Research Use Only

Formula: C₅H₄Br₂N₂
MW: 251.91 g/mol
MDL Number: MFCD00038041
CAS Number: 35486-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR