Methyl 4-bromo-1H-imidazole-2-carboxylate 95%

Supplier: Ambeed
Warning

A153911-100MG A153911-1G A153911-250MG
77010-926EA 101.85 USD
77010-926 77592-888 77592-890
Methyl 4-bromo-1H-imidazole-2-carboxylate 95%
Methyl 4-bromo-1H-imidazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O₂
MW: 205.00 g/mol
MDL Number: MFCD27922785
CAS Number: 1622843-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR