trans-Benzylideneacetone 98%

Supplier: Ambeed
Warning

Synonyms: (E)-4-Phenylbut-3-en-2-one, trans-4-Phenyl-3-buten-2-one

A153836-1000G A153836-100G A153836-500G
76997-452EA 68.96 USD
76997-452 76997-454 76997-456
trans-Benzylideneacetone 98%
trans-Benzylideneacetone

Precaución: for Research Use Only

Formula: C₁₀H₁₀O
MW: 146.19 g/mol
MDL Number: MFCD00008779
CAS Number: 1896-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR