4-Bromo-2-nitronaphthalen-1-amine 95%

Supplier: Ambeed
Warning

A153833-100MG A153833-250MG
76984-738EA 15 USD
76984-738 76984-740
4-Bromo-2-nitronaphthalen-1-amine 95%
4-Bromo-2-nitronaphthalen-1-amine

Precaución: for Research Use Only

Formula: C₁₀H₇BrN₂O₂
MDL Number: MFCD23703106
CAS Number: 90767-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR