2-Aminobenzothiazole 97%

Supplier: Ambeed
Warning

Synonyms: Benzothiazol-2-ylamine, Benzo[d]thiazol-2-amine, Cerium(III)-Ionophore II

A153771-1000G A153771-100G A153771-25G A153771-500G
76997-442EA 86 USD
76997-442 76997-444 76997-446 76997-448
2-Aminobenzothiazole 97%
2-Aminobenzothiazole

Precaución: for Research Use Only

Formula: C₇H₆N₂S
MDL Number: MFCD00005785
CAS Number: 136-95-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR