2-Amino-2-(4-fluorophenyl)ethanol 95%

Supplier: Ambeed
Warning

A153716-250MG A153716-1G A153716-50MG A153716-100MG A153716-5G
76997-440EA 25 USD
76997-440 77671-154 77671-156 77881-190 77881-191
2-Amino-2-(4-fluorophenyl)ethanol 95%
2-Amino-2-(4-fluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₁₀FNO
MDL Number: MFCD11213644
CAS Number: 140373-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR