Oxazole-5-methanol 97%

Supplier: Ambeed
Warning

Synonyms: Oxazol-5-ylmethanol, 5-(Hydroxymethyl)-1,3-oxazole

A153667-250MG A153667-5G A153667-1G
77010-922EA 50.93 USD
77010-922 77010-924 77683-289
Oxazole-5-methanol 97%
Oxazole-5-methanol

Precaución: For research use only.

Formula: C₄H₅NO₂
MW: 99.0880 g/mol
MDL Number: MFCD09027272
CAS Number: 127232-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR