Methyl 4-bromo-5-methylpicolinate 95%

Supplier: Ambeed
Warning

A153525-100MG A153525-1G A153525-250MG A153525-5G
76984-732EA 88.06 USD
76984-732 76984-734 76984-736 77683-288
Methyl 4-bromo-5-methylpicolinate 95%
Methyl 4-bromo-5-methylpicolinate

Precaución: For research use only.

Formula: C₈H₈BrNO₂
MW: 230.05 g/mol
MDL Number: MFCD18258411
CAS Number: 1256813-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR