Biphenyl-3,3',5,5'-tetracarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: [1,1'-Biphenyl]-3,3',5,5'-tetracarboxylic acid

A153483-1G A153483-5G A153483-250MG
76984-728EA 63.66 USD
76984-728 76984-730 77592-880
Biphenyl-3,3',5,5'-tetracarboxylic acid 95%
Biphenyl-3,3',5,5'-tetracarboxylic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₀O₈
MW: 330.25 g/mol
MDL Number: MFCD16621475
CAS Number: 4371-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR