Tozasertib 99+%

Supplier: Ambeed
Warning

Synonyms: Tozasertib, N-(4-((4-((5-Methyl-1H-pyrazol-3-yl)amino)-6-(4-methylpiperazin-1-yl)pyrimidin-2-yl)thio)phenyl)cyclopropanecarboxamide

A153482-1MG A153482-50MG
77028-272EA 7.43 USD
77028-272 77028-274
Tozasertib 99+%
Tozasertib

Precaución: for Research Use Only

Formula: C₂₃H₂₈N₈OS
MW: 464.59 g/mol
MDL Number: MFCD13185152
CAS Number: 639089-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR