2-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-3-oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-6-carboxylic acid

A153460-100MG A153460-1G A153460-250MG
76997-430EA 41.38 USD
76997-430 76997-432 76997-434
2-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylic acid 95%
2-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₄
MW: 207.19 g/mol
MDL Number: MFCD11207468
CAS Number: 1092352-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR