7-(Trifluoromethyl)-2,3-dihydro-1H-inden-1-one 95%

Supplier: Ambeed
Warning

A153327-1G A153327-250MG
76984-724EA 498.63 USD
76984-724 76984-726
7-(Trifluoromethyl)-2,3-dihydro-1H-inden-1-one 95%
7-(Trifluoromethyl)-2,3-dihydro-1H-inden-1-one

Precaución: for Research Use Only

Formula: C₁₀H₇F₃O
MW: 200.16 g/mol
MDL Number: MFCD11040251
CAS Number: 1003048-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR