N-(5-(((5-(tert-Butyl)oxazol-2-yl)methyl)thio)thiazol-2-yl)piperidine-4-carboxamide ≥99%

Supplier: Ambeed
Warning

Synonyms: SNS-032

A153280-100MG A153280-10MG A153280-1G A153280-1MG A153280-250MG A153280-50MG A153280-5MG A153280-5G
77028-258EA 203 USD
77028-258 77028-260 77028-262 77028-264 77028-266 77028-268 77028-270 77871-756
N-(5-(((5-(tert-Butyl)oxazol-2-yl)methyl)thio)thiazol-2-yl)piperidine-4-carboxamide ≥99%
N-(5-(((5-(tert-Butyl)oxazol-2-yl)methyl)thio)thiazol-2-yl)piperidine-4-carboxamide

Precaución: for Research Use Only

Formula: C₁₇H₂₄N₄O₂S₂
MDL Number: MFCD09833875
CAS Number: 345627-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR