5-Bromo-2-methyl-3-nitroaniline 97%

Supplier: Ambeed
Warning

A153015-10G A153015-1G A153015-25G A153015-5G
76984-704EA 49 USD
76984-704 76984-706 76984-708 76984-710
5-Bromo-2-methyl-3-nitroaniline 97%
5-Bromo-2-methyl-3-nitroaniline

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₂
MDL Number: MFCD07781362
CAS Number: 864550-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR