1-Isobutylpyrazole-4-carbaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 1-Isobutyl-1H-pyrazole-4-carbaldehyde

A152904-5G A152904-250MG
76997-428EA 228.1 USD
76997-428 77671-140
1-Isobutylpyrazole-4-carbaldehyde 95%
1-Isobutylpyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O
MW: 152.194 g/mol
MDL Number: MFCD08559037
CAS Number: 1006333-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR