2-Fluoro-4-chlorophenacyl bromide 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-1-(4-chloro-2-fluorophenyl)ethanone

A152874-250MG A152874-1G A152874-5G
77010-892EA 28.65 USD
77010-892 77592-864 77592-866
2-Fluoro-4-chlorophenacyl bromide 97%
2-Fluoro-4-chlorophenacyl bromide

Precaución: for Research Use Only

Formula: C₈H₅BrClFO
MW: 251.48 g/mol
MDL Number: MFCD13194236
CAS Number: 725743-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR