Methyl 2-amino-2-methylpropanoate 98%

Supplier: Ambeed
Danger

A152836-100MG A152836-1G A152836-250MG
77010-888EA 9 USD
77010-888 77010-890 77892-046
Methyl 2-amino-2-methylpropanoate 98%
Methyl 2-amino-2-methylpropanoate

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂
MDL Number: MFCD12407415
CAS Number: 13257-67-5
UN: 2924
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR