6-Methoxy-7-hydroxyquinazolin-4-one 97%

Supplier: Ambeed
Warning

A152814-100G A152814-10G A152814-1G A152814-250MG A152814-25G A152814-5G
76997-410EA 756.43 USD
76997-410 76997-412 76997-414 76997-416 76997-418 76997-420
6-Methoxy-7-hydroxyquinazolin-4-one 97%
6-Methoxy-7-hydroxyquinazolin-4-one

Precaución: for Research Use Only

Formula: C₉H₈N₂O₃
MW: 192.171 g/mol
MDL Number: MFCD08458441
CAS Number: 162012-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR