Methyl 1-methyl-1H-indazole-5-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 5-(Methoxycarbonyl)-1-methyl-1H-indazole

A152809-1G A152809-250MG A152809-25G
77028-238EA 60.48 USD
77028-238 77028-240 77028-242
Methyl 1-methyl-1H-indazole-5-carboxylate 97%
Methyl 1-methyl-1H-indazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.199 g/mol
MDL Number: MFCD11109401
CAS Number: 1092351-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR