3-(Dipropylamino)-4-methoxybenzenesulfonic acid 95%

Supplier: Ambeed
Warning

A152751-1G A152751-250MG A152751-5G
76997-404EA 71.09 USD
76997-404 76997-406 76997-408
3-(Dipropylamino)-4-methoxybenzenesulfonic acid 95%
3-(Dipropylamino)-4-methoxybenzenesulfonic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₁NO₄S
MW: 287.38 g/mol
MDL Number: MFCD09263742
CAS Number: 153086-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR