3-Chloro-4-isopropoxyphenylboronic acid 97%

Supplier: Ambeed
Warning

A152714-1G A152714-25G A152714-5G
77028-228EA 16 USD
77028-228 77028-230 77028-232
3-Chloro-4-isopropoxyphenylboronic acid 97%
3-Chloro-4-isopropoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BClO₃
MDL Number: MFCD04974065
CAS Number: 480438-56-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR