6-(3-Fluorophenyl)-3-pyridinecarbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 6-(3-Fluorophenyl)nicotinaldehyde

A152676-100MG A152676-1G A152676-250MG
77010-882EA 50 USD
77010-882 77010-884 77010-886
6-(3-Fluorophenyl)-3-pyridinecarbaldehyde 98%
6-(3-Fluorophenyl)-3-pyridinecarbaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₈FNO
MW: 201.2 g/mol
MDL Number: MFCD04115444
CAS Number: 898795-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR