Dichloro[1,1'-bis(dicyclohexylphosphino)ferrocene]palladium(II) 98+%

Supplier: Ambeed
Danger

Synonyms: PdCl{2}(dcypf), [1,1'-Bis(dicyclohexylphosphino)ferrocene]dichloropalladium(II), [1,1′-Bis(di-cyclohexylphosphino)ferrocene]dichloropalladium(II)

A152586-1G A152586-250MG
76984-694EA 59.42 USD
76984-694 77683-282
Dichloro[1,1'-bis(dicyclohexylphosphino)ferrocene]palladium(II) 98+%
Dichloro[1,1'-bis(dicyclohexylphosphino)ferrocene]palladium(II)

Precaución: For research use only.

Formula: C₃₄H₄₆Cl₂FeP₂Pd
MW: 749.85 g/mol
MDL Number: MFCD11656081
CAS Number: 917511-90-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR