(S)-Morpholine-3-carboxylic acid hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A152449-250MG
A152449-5G
A152449-1G
77010-878EA
9
USD
77010-878
77010-880
77890-003
(S)-Morpholine-3-carboxylic acid hydrochloride 97%
(S)-Morpholine-3-carboxylic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₀ClNO₃ |
MDL Number:
MFCD06809590 CAS Number: 1187929-04-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...