Methyl 2-(4-(benzyloxy)phenyl)acetate 97%

Supplier: Ambeed
Warning

A152445-10G A152445-1G A152445-250MG A152445-25G A152445-5G
77028-212EA 56.23 USD
77028-212 77028-214 77028-216 77028-218 77028-220
Methyl 2-(4-(benzyloxy)phenyl)acetate 97%
Methyl 2-(4-(benzyloxy)phenyl)acetate

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₃
MW: 256.29 g/mol
MDL Number: MFCD00157061
CAS Number: 68641-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR