3-Amino-5-ethyl-1H-pyrazole 97%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-5-ethylpyrazole

A152439-10G A152439-1G A152439-250MG A152439-25G A152439-5G
77010-868EA 199.45 USD
77010-868 77010-870 77010-872 77010-874 77010-876
3-Amino-5-ethyl-1H-pyrazole 97%
3-Amino-5-ethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₅H₉N₃
MW: 111.15 g/mol
MDL Number: MFCD08060982
CAS Number: 1904-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR