1-(1H-Pyrrolo[3,2-c]pyridin-3-yl)ethanone 98%

Supplier: Ambeed
Warning

A152426-1G A152426-250MG
77010-864EA 105.03 USD
77010-864 77010-866
1-(1H-Pyrrolo[3,2-c]pyridin-3-yl)ethanone 98%
1-(1H-Pyrrolo[3,2-c]pyridin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MW: 160.17 g/mol
MDL Number: MFCD11007991
CAS Number: 460053-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR