5-Methylthiophene-3-carbaldehyde 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Methyl-3-thiophenecarboxaldehyde
A152369-250MG
A152369-1G
76984-680EA
100
USD
76984-680
77889-999
5-Methylthiophene-3-carbaldehyde 95%
5-Methylthiophene-3-carbaldehyde
Precaución: for Research Use Only
Formula:
C₆H₆OS |
MDL Number:
MFCD07792887 CAS Number: 29421-72-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...