4'-Brom-3'-methyloacetophenone 95%

Supplier: Ambeed
Warning

A152368-100MG A152368-1G A152368-250MG A152368-5G
77010-850EA 20 USD
77010-850 77010-852 77010-854 77010-856
4'-Brom-3'-methyloacetophenone 95%
4'-Brom-3'-methyloacetophenone

Precaución: for Research Use Only

Formula: C₉H₉BrO
MDL Number: MFCD17169058
CAS Number: 50870-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR