Gatifloxacin sesquihydrate 95%

Supplier: Ambeed
Warning

Synonyms: 1-Cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid sesquihydrate

A152235-1MG A152235-250MG
76997-388EA 5.31 USD
76997-388 76997-390
Gatifloxacin sesquihydrate 95%
Gatifloxacin sesquihydrate

Precaución: for Research Use Only

Formula: C₃₈H₅₀F₂N₆O₁₁
MW: 804.83 g/mol
MDL Number: MFCD22373645
CAS Number: 180200-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR